2-Chloro-2'-deoxyadenosine, [8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT739-250UCI MT739-1MCI
101973-524EA 2700.38 USD
101973-524 101973-526
2-Chloro-2'-deoxyadenosine, [8-3H] ≥97% (by HPLC), MORpure™
2-Chloro-2'-deoxyadenosine, [8-3H]
CAS Number: 339527-32-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR