Ganciclovir, [8-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT725-500UCI MT725-1MCI
101973-502EA 4060.38 USD
101973-502 101973-504
Ganciclovir, [8-3H] ≥97%, MORpure™
Ganciclovir, [8-3H]
CAS Number: 315213-43-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR