7-Deazaadenosine [8-3H(N)] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT709-1MCI MT709-250UCI
101973-464EA 4125.77 USD
101973-464 101973-462
7-Deazaadenosine [8-3H(N)] ≥97%, MORpure™
7-Deazaadenosine [8-3H(N)]
CAS Number: 2086328-17-6
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR