Hypoxanthine, [2,8-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT700-1MCI MT700-5MCI
101973-438EA 3792.31 USD
101973-438 101973-440
Hypoxanthine, [2,8-3H] ≥97%, MORpure™
Hypoxanthine, [2,8-3H]
CAS Number: 33917-71-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR