2-Chloroadenosine, [8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT697-250UCI MT697-500UCI MT697-1MCI
101973-422EA 2700.38 USD
101973-422 101973-424 101973-426
2-Chloroadenosine, [8-3H] ≥97% (by HPLC), MORpure™
2-Chloroadenosine, [8-3H]
CAS Number: 99660-30-7
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR