Uridine-5'-monophosphate, diammonium salt, [5-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT691-250UCI MT691-1MCI
101973-410EA 2700.38 USD
101973-410 101973-412
Uridine-5'-monophosphate, diammonium salt, [5-3H] ≥97%, MORpure™
Uridine-5'-monophosphate, diammonium salt, [5-3H]
CAS Number: 339340-55-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR