5-Fluorouracil, [6-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT686-250UCI MT686-1MCI
101973-402EA 2700.38 USD
101973-402 101973-404
5-Fluorouracil, [6-3H] ≥97% (by HPLC), MORpure™
5-Fluorouracil, [6-3H]
CAS Number: 23935-92-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR