L-Leucine, [4,5-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT672-250UCI MT672-1MCI MT672-5MCI
101973-350EA 477.31 USD
101973-350 101973-352 101973-354
L-Leucine, [4,5-3H] ≥97%, MORpure™
L-Leucine, [4,5-3H]
CAS Number: 2087903-59-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR