Orotic acid, [5-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT668-250UCI MT668-1MCI
101973-342EA 2700.38 USD
101973-342 101973-344
Orotic acid, [5-3H] ≥97% (by HPLC), MORpure™
Orotic acid, [5-3H]
CAS Number: 24465-39-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR