Uracil, [6-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT656-250UCI MT656-1MCI MT656-5MCI
101973-308EA 2700.38 USD
101973-308 101973-310 101973-312
Uracil, [6-3H] ≥97% (by HPLC), MORpure™
Uracil, [6-3H]
CAS Number: 6165-70-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR