2'-Deoxyuridine, [6-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT649-250UCI MT649-1MCI
101973-296EA 2700.38 USD
101973-296 101973-298
2'-Deoxyuridine, [6-3H] ≥97%, MORpure™
2'-Deoxyuridine, [6-3H]
CAS Number: 65370-72-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR