5-Bromo-2'-deoxyuridine, [6-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT637-250UCI MT637-1MCI
101973-266EA 2700.38 USD
101973-266 101973-268
5-Bromo-2'-deoxyuridine, [6-3H] ≥97%, MORpure™
5-Bromo-2'-deoxyuridine, [6-3H]
CAS Number: 69655-10-3
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR