Aflatoxin-B2, [8,9-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT636-250UCI MT636-1MCI
101973-262EA 2700.38 USD
101973-262 101973-264
Aflatoxin-B2, [8,9-3H(N)] ≥97% (by HPLC), MORpure™
Aflatoxin-B2, [8,9-3H(N)]
CAS Number: 90704-23-7
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR