Cytosine-β-D-arabinofuranoside, [5-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT631-250UCI MT631-1MCI
101973-248EA 1601.92 USD
101973-248 101973-250
Cytosine-β-D-arabinofuranoside, [5-3H(N)] ≥97% (by HPLC), MORpure™
Cytosine-β-D-arabinofuranoside, [5-3H(N)]
CAS Number: 60299-13-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR