2'-Deoxyuridine-5'-triphosphate, tetraammonium salt, [5-3H] ≥96% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT621-250UCI MT621-1MCI MT621-5MCI
101973-234EA 2700.38 USD
101973-234 101973-236 101973-238
2'-Deoxyuridine-5'-triphosphate, tetraammonium salt, [5-3H] ≥96% (by HPLC)
2'-Deoxyuridine-5'-triphosphate, tetraammonium salt, [5-3H]
CAS Number: 339163-21-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR