Adenosine-5'-monophosphate, diammonium salt, [2,8-3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT618-250UCI MT618-1MCI MT618-5MCI
101973-222EA 2700.38 USD
101973-222 101973-224 101973-226
Adenosine-5'-monophosphate, diammonium salt, [2,8-3H] ≥95% (by HPLC)
Adenosine-5'-monophosphate, diammonium salt, [2,8-3H]
CAS Number: 339163-16-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR