Thymine, [methyl-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT609-1MCI MT609-5MCI
101973-192EA 3046.92 USD
101973-192 101973-194
Thymine, [methyl-3H] ≥97%, MORpure™
Thymine, [methyl-3H]
CAS Number: 78298-96-1
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR