Thymidine, [methyl-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT6038-250UCI MT6038-1MCI MT6038-5MCI
101973-160EA 536.15 USD
101973-160 101973-162 101973-164
Thymidine, [methyl-3H] ≥97%, MORpure™
Thymidine, [methyl-3H]
CAS Number: 52497-68-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR