Thymidine, [methyl-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT6032-250UCI MT6032-1MCI MT6032-5MCI
101973-130EA 536.15 USD
101973-130 101973-132 101973-134
Thymidine, [methyl-3H] ≥97% (by HPLC), MORpure™
Thymidine, [methyl-3H]
CAS Number: 52497-68-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR