2',3'-Dideoxycytidine, [2',3'-3H(N)] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT586-50UCI MT586-250UCI
101973-100EA 1601.92 USD
101973-100 101973-102
2',3'-Dideoxycytidine, [2',3'-3H(N)] ≥97%, MORpure™
2',3'-Dideoxycytidine, [2',3'-3H(N)]
CAS Number: 2086328-30-3
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR