5'-N-Ethylcarboxamidoadenosine [adenine-2,8-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT581-250UCI MT581-1MCI
101973-090EA 2700.38 USD
101973-090 101973-092
5'-N-Ethylcarboxamidoadenosine [adenine-2,8-3H] ≥97%, MORpure™
5'-N-Ethylcarboxamidoadenosine [adenine-2,8-3H]
CAS Number: 151362-68-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR