Methylamine hydrochloride, [methyl-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT545-50UCI MT545-250UCI MT545-1MCI
101973-040EA 1601.92 USD
101973-040 101973-042 101973-044
Methylamine hydrochloride, [methyl-3H] ≥97% (by HPLC), MORpure™
Methylamine hydrochloride, [methyl-3H]
CAS Number: 127117-27-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR