Mitoxantrone, [3H] ≥90%

Supplier: MORAVEK BIOCHEMICALS MS

MT543-50UCI MT543-250UCI
101973-036EA 1896.15 USD
101973-036 101973-038
Mitoxantrone, [3H] ≥90%
Mitoxantrone, [3H]
CAS Number: 316351-61-8
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR