Guanosine, [8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT514-250UCI MT514-1MCI MT514-5MCI
101972-974EA 2700.38 USD
101972-974 101972-976 101972-978
Guanosine, [8-3H] ≥97% (by HPLC), MORpure™
Guanosine, [8-3H]
CAS Number: 24613-04-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR