Uracil, [5,6-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT512-250UCI MT512-1MCI
101972-962EA 2576.15 USD
101972-962 101972-964 101972-966
Uracil, [5,6-3H] ≥97%, MORpure™
Uracil, [5,6-3H]
CAS Number: 35978-19-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR