Acycloguanosine, [8-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT508-1MCI MT508-250UCI MT508-500UCI
101972-954EA 4537.69 USD
101972-954 101972-950 101972-952
Acycloguanosine, [8-3H] ≥97%, MORpure™
Acycloguanosine, [8-3H]
CAS Number: 116981-61-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR