Ciclopirox, [³H]- 1.0 mCi/ml in ethanol ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1971-50UCI MT1971-250UCI
101972-904EA 1601.92 USD
101972-904 101972-906
Ciclopirox, [³H]- 1.0 mCi/ml in ethanol ≥97% (by HPLC), MORpure™
Ciclopirox, [³H]-

MORpure™ HPLC purified, Delivered ≥ 97.0% pure with dated HPLC radiochromatogram

Storage Temperature: Freezer
CAS Number: 29342-05-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR