4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-(5-(p-Tolyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulphonamide, Celecoxib

MT1965-50UCI MT1965-250UCI
101972-886EA 1601.92 USD
101972-886 101972-888
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
Formula: C₁₇H₁₄F₃N₃O₂S
MW: 381.38 g/mol
MDL Number: MFCD00941298
CAS Number: 169590-42-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR