Entecavir, [3H]

Supplier: MORAVEK BIOCHEMICALS MS

MT1951-50UCI MT1951-250UCI
101972-844EA 3092.69 USD
101972-844 101972-846
Entecavir, [3H]
Entecavir, [3H]
CAS Number: 142217-69-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR