Nicotinamide, [6-3H(N)] ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

MT1937-250UCI MT1937-1MCI
101972-832EA 2700.38 USD
101972-832 101972-834
Nicotinamide, [6-3H(N)] ≥95%
Nicotinamide, [6-3H(N)]
CAS Number: 2086336-93-6
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR