Amodiaquin, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1928-50UCI MT1928-250UCI
101972-812EA 1896.15 USD
101972-812 101972-814
Amodiaquin, [3H] ≥97% (by HPLC), MORpure™
Amodiaquin, [3H]
CAS Number: 86-42-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR