Dihydroantillatoxin, [3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT1924-50UCI MT1924-250UCI
101972-796EA 1601.92 USD
101972-796 101972-798
Dihydroantillatoxin, [3H] ≥96%
Dihydroantillatoxin, [3H]
CAS Number: 2086327-53-7
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR