L-Phenylalanine, [ring-2,4-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1916-250UCI MT1916-1MCI
101972-790EA 2700.38 USD
101972-790 101972-792
L-Phenylalanine, [ring-2,4-3H] ≥97%, MORpure™
L-Phenylalanine, [ring-2,4-3H]
CAS Number: 316366-07-1
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR