Domperidone

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 5-Chloro-1-[1-[3-(2-oxo-1-benzimidazolyl)propyl]-4-piperidyl]benzimidazol-2-one

MT1857-100UCI MT1857-250UCI MT1857-1MCI
101972-648EA 2281.92 USD
101972-648 101972-650 101972-652
Domperidone
Domperidone
Formula: C₂₂H₂₄ClN₅O₂
MW: 425.92 g/mol
MDL Number: MFCD00069256
CAS Number: 57808-66-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR