Stearoyl coenzyme A, [stearoyl-9,10-3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1849-50UCI MT1849-250UCI
101972-636EA 2085.77 USD
101972-636 101972-638
Stearoyl coenzyme A, [stearoyl-9,10-3H] ≥95% (by HPLC)
Stearoyl coenzyme A, [stearoyl-9,10-3H]
CAS Number: 313949-40-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR