Triamcinolone acetonide
Supplier: MORAVEK BIOCHEMICALS MS
|
|
MT1754-50UCI
MT1754-250UCI
101972-558EA
1601.92
USD
101972-558
101972-560
Triamcinolone acetonide
Triamcinolone acetonide
Formula:
C₂₄H₃₁FO₆ |
MDL Number:
MFCD00056834 CAS Number: 76-25-5 UN: 2910 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...