Tenofovir disoproxil fumarate

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate fumarate

MT1725-50UCI MT1725-250UCI
101972-496EA 2497.69 USD
101972-496 101972-498
Tenofovir disoproxil fumarate
Tenofovir disoproxil fumarate
Formula: C₁₉H₃₀N₅O₁₀P·C₄H₄O₄
MW: 635.5 g/mol
MDL Number: MFCD08141829
CAS Number: 202138-50-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR