17-Allylaminogeldanamycin [allylamino-2,3-3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1717-50UCI MT1717-250UCI
101972-480EA 1601.92 USD
101972-480 101972-482
17-Allylaminogeldanamycin [allylamino-2,3-3H] ≥95% (by HPLC)
17-Allylaminogeldanamycin [allylamino-2,3-3H]
CAS Number: 2086333-30-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR