5-Aza-2'-deoxycytidine, [6-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1676-250UCI MT1676-500UCI
101972-416EA 2700.38 USD
101972-416 101972-418
5-Aza-2'-deoxycytidine, [6-3H] ≥97%, MORpure™
5-Aza-2'-deoxycytidine, [6-3H]
CAS Number: 1140527-39-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR