L-Valine, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1654-250UCI MT1654-1MCI
101972-398EA 2700.38 USD
101972-398 101972-400
L-Valine, [3H] ≥97% (by HPLC), MORpure™
L-Valine, [3H]
CAS Number: 53521-25-8
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR