Oleoyl coenzyme A, [oleoyl-9,10-3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1649-10UCI MT1649-50UCI
101972-394EA 1222.69 USD
101972-394 101972-396
Oleoyl coenzyme A, [oleoyl-9,10-3H] ≥95% (by HPLC)
Oleoyl coenzyme A, [oleoyl-9,10-3H]
CAS Number: 2089214-47-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR