Arachidonyl coenzyme A, [arachidonyl-5,6,8,9,11,12,14,15-3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1641-50UCI MT1641-250UCI
101972-382EA 2569.62 USD
101972-382 101972-384
Arachidonyl coenzyme A, [arachidonyl-5,6,8,9,11,12,14,15-3H] ≥95% (by HPLC)
Arachidonyl coenzyme A, [arachidonyl-5,6,8,9,11,12,14,15-3H]
CAS Number: 315178-67-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR