Cytidine-5'-diphosphocholine, disodium salt [5-3H] ≥96% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1609-10UCI MT1609-50UCI MT1609-250UCI
101972-326EA 1418.85 USD
101972-326 101972-328 101972-330
Cytidine-5'-diphosphocholine, disodium salt [5-3H] ≥96% (by HPLC)
Cytidine-5'-diphosphocholine, disodium salt [5-3H]
CAS Number: 112820-38-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR