Resveratrol, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1606-50UCI MT1606-250UCI MT1606-1MCI
101972-316EA 1601.92 USD
101972-316 101972-318 101972-320
Resveratrol, [3H] ≥97% (by HPLC), MORpure™
Resveratrol, [3H]
CAS Number: 339155-28-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR