Ribavirin-5'-triphosphate, tetraammonium salt, [3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1593-50UCI MT1593-250UCI
101972-296EA 1601.92 USD
101972-296 101972-298
Ribavirin-5'-triphosphate, tetraammonium salt, [3H] ≥95% (by HPLC)
Ribavirin-5'-triphosphate, tetraammonium salt, [3H]
CAS Number: 339155-22-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR