rac-2,6,-Diaminopimelic acid, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1556-250UCI MT1556-1MCI
101972-236EA 2700.38 USD
101972-236 101972-238
rac-2,6,-Diaminopimelic acid, [3H] ≥97% (by HPLC), MORpure™
rac-2,6,-Diaminopimelic acid, [3H]
CAS Number: 50298-83-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR