Bisphenol A, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1551-250UCI MT1551-1MCI
101972-232EA 2700.38 USD
101972-232 101972-234
Bisphenol A, [3H] ≥97% (by HPLC), MORpure™
Bisphenol A, [3H]
CAS Number: 337963-21-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR