Succinic acid, [2,3-3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT1528-250UCI MT1528-1MCI
101972-200EA 2700.38 USD
101972-200 101972-202
Succinic acid, [2,3-3H] ≥96%
Succinic acid, [2,3-3H]
CAS Number: 3073-84-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR