Flutamide, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1520-250UCI MT1520-1MCI
101972-178EA 2700.38 USD
101972-178 101972-180
Flutamide, [3H] ≥97% (by HPLC), MORpure™
Flutamide, [3H]
CAS Number: 2086731-38-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR