Clotrimazole, [3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT1519-250UCI MT1519-1MCI
101972-174EA 2700.38 USD
101972-174 101972-176
Clotrimazole, [3H] ≥96%
Clotrimazole, [3H]
CAS Number: 337962-97-7
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR